Would you like to log in again?
Clorox® Bleach Cream Cleanser, Fresh Scent, Essendant
Supplier: Essendant
Clorox®
CLO30613
76500-230CT 84.71 USD InStock
76500-230
Clorox® Bleach Cream Cleanser, Fresh Scent, Essendant
Cleaning Agents Detergents and Surface Cleansers
With the power of Clorox® bleach, Clorox® Bleach Cream Cleanser removes stubborn stains and leaves surfaces clean and sanitary.
  • Fast, strong cleaning power to tackle the toughest stains and clean any surface
  • Sodium hypochlorite solution provides tough cleaning power that’s gentle on surfaces
  • Color-enriched formula shows where it's been applied for complete coverage and easy cleaning
  • Easy to rinse off and gentle on surfaces
  • Leaves behind a bright shine and fresh scent
Easy to use and rinse, yet gentle on surfaces. With 1.3% sodium hypochlorite, it provides fast, strong cleaning. From Clorox Commercial Solutions®, this cleaner is tested for compatibility with most surfaces including sinks and tubs, showers, toilets, kitchen countertops, stainless steel appliances, and more. Ideal for use in healthcare settings.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty