Would you like to log in again?
Oxivir® Five 16 One-Step Disinfectant Cleaner, Essendant
Supplier: Essendant
DVO4963314
76500-298CT 328.03 USD InStock
76500-298
Oxivir® Five 16 One-Step Disinfectant Cleaner, Essendant
Cleaning Agents Disinfectants
Proprietary Accelerated Hydrogen Peroxide (AHP®) based disinfectant-cleaner delivers fast, effective cleaning performance.
  • Hospital-grade disinfectant-cleaner
  • Based on proprietary, Accelerated Hydrogen Peroxide (AHP®) technology
  • Disinfects in 5 minutes at 1:16 dilution ratios
  • Excellent surface safety
  • HMIS ratings of zero
At a 1:16 dilution ratio, product disinfects hard, non-porous surfaces in five minutes. Use as a virucide, bactericide, fungicide, mildewcide, and a non-food contact sanitizer. Kills MRSA, norovirus, and canine parvovirus. Suitable for use on ceramic, fiberglass, glass, laminates, metal, plastic, sealed stone, and vinyl. Chemical compound: 2-Hydroxybenzoic Acid; Phosphoric Acid; Hydrogen Peroxide.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty