Would you like to log in again?
Comet® Disinfecting-Sanitizing Bathroom Cleaner, Essendant
Supplier: Essendant
Comet®
PGC22570CT
76500-312CT 132.99 USD InStock
76500-312
Comet® Disinfecting-Sanitizing Bathroom Cleaner, Essendant
Cleaning Agents Disinfectants
Comet® Disinfecting-Sanitizing Bathroom Cleaner is a powerful bathroom disinfectant cleaner that cuts through tough soap scum, leaving surfaces clean and disinfected (when used per disinfecting label instructions).
  • Combines the power of a heavy-duty restroom cleaner and surface deodorizer with a disinfectant
  • Non-abrasive bathroom sanitizer spray cleans greasy soap scum with 50% fewer strokes, saving time and labor
  • Included on the EPA List N: disinfectants for use against the novel coronavirus SARS-CoV-2
  • Cleans and disinfects in one step when used per disinfecting label instructions
This patented no scrub bathroom cleaner formula uniquely penetrates tough bathroom soils with little effort. It’s non-abrasive, so it safely cleans porcelain, fiberglass, ceramic, Formica, enamel, chrome, stainless steel, and synthetic marble. With no rinsing required, just spray it on and wipe the surface clean. This disinfecting bathroom cleaner deodorizes, leaving the restroom smelling clean and fresh.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty