Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Dawn Pro Heavy-Duty Degreaser, Essendant
Supplier: Essendant
Dawn®
PGC04852
76500-334CT 130.95 USD InStock
76500-334
Dawn Pro Heavy-Duty Degreaser, Essendant
Cleaning Agents Detergents and Surface Cleansers
Cuts through tough jobs with the grease-cutting power of Dawn.
  • Great for tough build-up on range hoods, ovens, walls, floors, and drive-thru pads
  • Full strength for greasy soils
  • Commercial degreaser can be used on food-contact surfaces when followed by rinsing with water
  • Formulated for employee safety and is phosphate-free, non-caustic, and non-flammable
This industrial degreaser is designed to clean the toughest grease problems. It can be used full strength as a multi-surface cleaner, and it is safe for use on food-contact surfaces when followed by rinsing with water. Excellent for cleaning corral areas and dumpsters and can even be used for cleaning heavy crank case oil from concrete drive-thru pads and sidewalks. It's also designed to clean waxed tile floors without damaging the wax, when properly diluted, and can spot-clean where heavy grease buildup exists.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty