Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Tide Pro Floor and All-Purpose Cleaner, Essendant
Supplier: Essendant
Tide®
PGC02364
76500-358EA 238.75 USD InStock
76500-358
Tide Pro Floor and All-Purpose Cleaner, Essendant
Cleaning Agents Detergents and Surface Cleansers
Tide all purpose floor cleaner is an economical and versatile cleaning solution that removes tough grease and lifts away tracked-in dirt.
  • High levels of surfactant and builders combined to cut through tough grease
  • Easy-rinse powder formula is ideal for quarry tile
  • Can be used with chlorine bleach
  • Contains no phosphates
This multi surface cleaner can be used with chlorine bleach and works great in all areas of the kitchen. It can be used on floors, walls, pots and pans, quarry tiles, unwaxed floors, exhaust filter screens and hoods, cooler ceilings, walls and racks, mop heads, and outside areas such as concrete and dumpsters. It is formulated for industrial use. Unscented.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty