Would you like to log in again?
Clorox® VersaSure™ Cleaner Disinfectant Wipes, Essendant
Supplier: Essendant
Clorox®
CLO31757
76500-374CT 85.45 USD InStock
76500-374
Clorox® VersaSure™ Cleaner Disinfectant Wipes, Essendant
Wipes Disposable Wipes and Towels Disinfectant Wipes
Clorox® Healthcare VersaSure™ Cleaner Disinfectant Wipes kill microorganisms quickly on a broad range of surfaces. These strong and durable alcohol-free wipes are a one-step solution to cleaning and disinfecting.
  • Quaternary ammonium disinfecting wipes
  • Wipes are noncorrosive and contain no added fragrances, chemical odors, fumes, or bleach
  • Disinfect most bacteria and viruses in as little as 30 seconds
VersaSure™ wipes are EPA-registered to kill pathogens on hard surfaces in as fast as 30 seconds for most bacteria and viruses. Designed for patient and staff comfort, these wipes are formulated with no harsh chemical odors, fumes, or added fragrances. Intended for use in commercial settings. Wipes are suitable for use on aluminum, baked enamel, brass, chrome, copper, corian, finished wood, formica, galvanized steel, glass, glazed porcelain, laminate, linoleum, marble, metal, naugahyde, painted metal, painted wood, plastic, plastic laminate, plexiglass, porcelain enamel, rubber, sealed fiberglass, sealed granite, sealed wood, silver, stainless steel, synthetic marble, treated wood, vinyl tile, and wood.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty