Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Clorox® Fuzion Cleaner Disinfectant, Unscented, Essendant
Supplier: Essendant
Clorox®
CLO31478
76500-378CT 246.23 USD InStock
76500-378
Clorox® Fuzion Cleaner Disinfectant, Unscented, Essendant
Cleaning Agents Disinfectants
This one-step, fast acting, ready-to-use cleaning spray combines the trusted efficacy of bleach with excellent surface compatibility and low odor.
  • Multi-surface cleaner with bleach is fast acting and ready to use
  • Powerful bleach cleaner contains a neutralizer to significantly reduce residue and reduce lingering bleach odor
  • Broad surface compatibility on surface materials
  • Covers twice as much area with each spray, when compared with traditional swirl nozzles.
  • No PPE required
This next-generation bleach product is EPA-registered to kill 53 microorganisms with a 2 minute kill time for C. difficile spores, the fastest kill time available. It offers a 1-minute kill time for a broad range of bacteria, viruses, and fungi, including Norovirus, MRSA, TB, and VRE. Fuzion Cleaner Disinfectant features a revolutionary dual chamber technology that combines bleach solution and neutralizing solution at the nozzle. The neutralizer in the formula breaks down the bleach to significantly reduce residue and eliminate lingering odor. The ergonomic bottle shape and new trigger technology delivers even surface coverage, clings to vertical surfaces with minimal dripping, and covers twice as much area with each spray. Suitable for use on most surfaces, it does not cause long-term damage to equipment like staining, cracking, or corroding. Intended for use in multiple commercial settings including healthcare, schools, athletic facilities, and long-term care facilities.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty