Would you like to log in again?
Comet® Liquid Cleaner with Bleach, Essendant
Supplier: Essendant
Comet®
PGC02291CT
76500-390CT 101.7 USD InStock
76500-390
Comet® Liquid Cleaner with Bleach, Essendant
Cleaning Agents Disinfectants
A unique two-in-one, all-purpose cleaner that combines the cleaning power of heavy-duty detergents with the stain-removal power of bleach.
  • Bleaches tough stains and cuts tough grease
  • Liquid chlorine bleach ingredient provides a wealth of well-known benefits
  • Removes tough kitchen soils such as tacky grease on range hoods
  • Bleaches food stains on countertops, tables, and cutting boards
Contains patented cleaning technologies and no abrasives to clean the full range of tough kitchen and restroom soils like tracked-in dirt and scuffmarks. Task areas include customer and employee restroom surfaces, kitchens, the inside and outside of plastic trash containers, and greasy equipment containers. Suitable for use on ceramic, fiberglass, plastic, porcelain, stainless steel, and tile.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty