Would you like to log in again?
Boardwalk® Quick Change Wooden Mop Handle, Essendant
Supplier: Essendant
Boardwalk®
BWK605
76500-432EA 20.93 USD InStock
76500-432
Boardwalk® Quick Change Wooden Mop Handle, Essendant
Mops
Specifically designed to fit mop heads sized #20 and above.
  • Lacquered wood handle with natural finish
  • Handle diameter: 1.13"
  • Handle length: 54"
Easy loading wooden mop handle allows quick change with metal framed mop heads.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty