Would you like to log in again?
BS100MONITORHFHEAT-SEALEDCN045UM HF PK48
Supplier: SARTORIUS CORPORATION EDI
Monitor, Biosart 100, cellulose nitrate, Heat sealed, membrane fixed, white filter black grids, membrane fixed, 100ml, 47mm, high flow 0.45um pore size, sterile packaged, designed for microbiological testing of pharmaceuticals and cosmetics, beverages and foods, water and other liquids
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
BS100MONITORHFHEAT-SEALEDCN045UM HF PK48 76500-736 1640347H6VWMK Pack of 48 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty