Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

BioSart® 100 Monitors, Sartorius

Supplier: Sartorius

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

BioSart®
164034704K 164034704VK 164014706K 1640147H6K 164034706VK 164014707K 164014707ACK 164014706VK 164034706K 164044706K 164044706K 164034706ACK 164014707ACK 164034706K 164034704K 16401-47-06--ACK 164024706K 1640147H6K 164024706ACK 164034704VK 164014706K 164034706VK 164014707K 1640147H6VK 1640147H6VK 164014706ACK 164014706VK 164034706ACK
76500-710PK 154.07 USD InStock
76500-710 76500-732 76500-730 CA76517-732 CA76517-754 CA76517-752 CA76517-730 CA76517-750 76500-708 76500-706 76500-728 CA76517-738 76500-726 76500-724 CA76517-756 CA76517-734 76500-720 CA76517-742 102517-206 CA76517-762 CA76517-740 102517-208 76500-718 76500-716 76500-738 CA76517-748 76500-714 CA76517-744
BioSart® 100 Monitors, Sartorius
Filters Membrane Filter Units
BioSart® 100 monitors have been specifically designed for the detection and enumeration of microorganisms in beverages, water, pharmaceuticals, cosmetics, foods and other liquids. These sterile devices with PS housing and incorporated membrane filter and cellulose pad are ready to use.

  • High flow rate and total throughput
  • Without any hydrophobic adhesive areas
  • High flow membranes allow shorter filtration times due to 30% higher flow rates
  • Funnel with 10 ml graduations
  • 6.5×1.5 mm outlet

After filtration of the sample, just add selected BioSart® media ampoule to the monitor and remove then the 100 ml funnel to convert the monitor into a petri dish eliminating the need for membrane manipulation.

These units are designed to be placed onto the bases of a vacuum manifold, eliminating the cleaning and sterilization required of reusable funnels.

Certifications: Certified according to the requirements of the International Standard ISO 9001. Lot certificates for recovery rate, sterility and specifications.

Packaging: 48 units per box. 3 different packaging types available:
- Single sterile packed: 12 single sterile monitors in 1 tray, 1 box includes 4 of these trays. Suitable for more critical applications like in the Pharma industry.
- Sterile packed in one tray: 12 monitors sterile packed in 1 tray, 1 box includes 4 of these trays.
- Sterile bulk packed in one box: 48 sterile monitors without extra packaging in 2 layers in 1 box, the box is wrapped in a thin foil. Suitable for less critical applications.

Order Now
 

ORDER

ACCESSORIES

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty