Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Forceps for Membrane Filters, Sartorius
Supplier: Sartorius
16625 16625
76500-760EA 50.02 USD InStock
CA76517-784 76500-760
Forceps for Membrane Filters, Sartorius
Forceps
These filter forceps are made of high grade stainless steel.
  • Autoclavable, can be flamed
  • With smooth, flat tips for a careful, firm hold of the membrane filter
Membrane filters should only be handled with suitable tweezers in order to avoid contamination which can result from hand contact.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty