Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.

Microsart® Vacuum Filtration Manifolds, Sartorius

Supplier: Sartorius
Microsart®
168M2BS100 168M1SS100 168M2MS 168M1SS500 168M6SS100 168M6SS500 168M6BS100 168M6MS 168M3SS100 168M1SS100 168M6BS250 168M3BS100 168M3BS100 168M3SS100 168M1MS 168M2BS100 168M6SS500 168M6BS100 168M6SS100 168M3BS250 168M3SS500 168M2MS 168M3SS500 168M6MS 168M3MS 168M3MS 168M6BS250 168M3BS250 168M1MS 168M1SS500
76500-798EA 3942.66 USD InStock
76500-798 76500-796 76500-794 76500-792 CA76517-818 76500-790 CA76517-830 76500-808 76500-806 CA76517-816 76500-804 CA76517-814 76500-802 CA76517-812 76500-800 76500-788 CA76517-832 CA76517-810 76500-786 76500-784 CA76517-808 CA76517-820 CA76517-806 CA76517-828 CA76517-826 CA76517-804 CA76517-824 76500-812 CA76517-822 76500-810
Microsart® Vacuum Filtration Manifolds, Sartorius
Manifolds
Microsart® Manifolds accurately detect and quantify microorganisms in liquid samples. Simply select the proper manifold for your individual microbiological testing procedure, choose between reusable and single-use funnels or filtration units, and then decide between different sizes of manifolds and enjoy the manifold that adapts to your needs.

  • 100% stainless steel for minimised cleaning effort and full autoclavability without taking apart
  • Sterile venting in critical applications reduces risk of secondary contamination
  • Quick connection adapters for fast connection of tubing, manifolds or end caps
  • Rubber feet and low working height for safe and stable ergonomic working procedures

Microsart® Manifold for Microsart®@filter and Microsart Funnels (pharmaceutical applications, cosmetics: Manifold for single-use filtration units)
• Add single sterile consumables: Microsart®@filter or Microsart Funnels
• For minimal risk of cross contamination by integrated venting and by touch-free membrane transfer when combining Microsart@filter use with Microsart@media plates

Microsart® Manifold for Biosart® 100 monitors (beverage and environmental water testing: Manifold for single-use filtration units)
• Add consumables: Biosart® 100 Monitor and Biosart® 100 media
• For minimal risk of cross contamination by integrated venting and by touch-free inoculation
• Media conforms to international regulations

Microsart® Manifold for Biosart® 250 funnels (optional beverage and environmental: Manifold for autoclavable funnels)
• Add autoclavable Biosart® 250 Funnels and Membrane filter
• Flexible easy quick connect set-up to adapt to variable numbers of tests per day
• Easy cleaning procedure, fully autoclavable manifold, no material mix

Microsart® Manifold with stainless steel funnels (optional beverage and environmental: Manifold with stainless steel funnels)
• Add membrane filter
• Flexible easy quick connect set-up to adapt to variable numbers of tests per day
• Easy cleaning by no dismantling, fully autoclavable manifold, no material mix
• Flammable stainless steel funnels (100 or 500 ml)

Delivery information: Manifolds include filter base support with sinter frit, blind coupling, 20 cm tubing and venting filter.

Order Now
 

ORDER

ACCESSORIES

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty