Would you like to log in again?
MICROSART ADDFILTER 250PC0.2UMBAG PK48
Supplier: SARTORIUS CORPORATION EDI
Filter, Microsart Add 250, Material: PC, Pore size: 0.2um bag, For use with Microsart Mani-folds funnel, a filter base, and a gridded membrane filter in one unit. For the detection and enumeration of microorganisms in pharmaceuticals, cosmetics, foods, beverages, water, and other liquids.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
MICROSART ADDFILTER 250PC0.2UMBAG PK48 76500-894 16D062507BK Pack of 48 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty