Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Filter Holder, Sartorius
Supplier: Sartorius
16213 16213
76501-008EA 667.09 USD InStock
CA76518-040 76501-008
Filter Holder, Sartorius
Filter Holders
Stainless steel closed filter holder for microbiological, analytical sampling and testing of gas in pressurized lines.
  • Streamlined design with ideal flow path geometry
  • Can be sterilized in a hot air oven
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty