Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Disposable Forceps, Burkle
Supplier: Bürkle
5378-1051 5378-1052 5378-1050 5378-1041 5378-0042 5378-1040 5378-1042 5378-0040 5378-0041
76501-432PK 58.93 USD InStock
76501-432 76501-434 76501-422 76501-430 76501-428 76501-436 76501-424 76501-438 76501-426
Disposable Forceps, Burkle
Forceps
These disposable forceps are suitable for precise lab work.
  • PS, white or blue
  • Individually packaged and optionally sterilized
  • For single use
  • Clean room manufactured (clean room class 7)
  • Overall length 130 mm (5.12")
They are ideal for grasping, lifting and moving small objects, for example fine tissue, small items or solid samples.
Certifications: Complies with EU food requirements and FDA regulations.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty