Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Easy Sampler, Burkle

Supplier: Bürkle

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

5360-1000 5360-0600
76501-530EA 1137.01 USD InStock
76501-530 76501-528
Easy Sampler, Burkle
Samplers Solid Samplers
The Easy Sampler is designed to quickly and easily take multiple samples from bulk goods.

  • Stainless steel AISI 316 L (1.4404)
  • For sample openings from 52 mm (2.05") diameter

Emptying has been greatly simplified and in this model takes place via the tip. The sample can be emptied directly into the sample container without having to disassemble the device, in order to obtain a mixed sample for instance. The bulk material must be free-flowing or self-flowing.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty