Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Azure™ Imaging Systems, Azure Biosystems
Supplier: Azure Biosystems
AZI300-01 AZI500-01 AZI30Q AZI200-01 AZI600-01 AZI50Q AZI400-01
76501-640EA 42574.32 USD InStock
76501-640 76501-642 76501-638 76501-644 76353-894 76501-636 76353-896
Azure™ Imaging Systems, Azure Biosystems
Photodocumentation Systems
Innovative western blot and gel documentation systems designed to be easy to use lab workhorses. All our systems deliver highly reproducible, quantitative data.
  • Designed for flexible Western blot applications
  • A wide range of applications with multiple light sources and filters
The 200 gel imaging workstation can image multiple types of gels. The darkroom replacer, model 300, features fast chemiluminescent detection with no film or darkroom needed. It can be upgraded to any of the ascending Azure Imager models (400, 500 or 600) with a simple field upgrade.
The visible fluorescent Western imaging system, model 400, features a three-channel RGB excitation for Cy5/Cy3/Cy2 Western applications. It also features fast chemiluminescent detection with no film or darkroom needed. It can be upgraded to an IR imaging system (model 600) with a simple field upgrade.
The infrared imaging system, model 500, features two-channel IR direct detection with laser diodes at 685 nm and 785 nm. Narrow-band excitation enables NIR applications with better sensitivity than LEDs or white light sources. It can image multiple types of gels and blots. It also features fast chemiluminescent detection with no film or darkroom needed. It can be upgraded to the model 600 with a simple field upgrade.The included UV and white light transillumination allows fluorescent and white-light imaging of gels, including nucleotide gels stained with ethidium bromide and protein gels stained with Coomassie blue or silver stain. It provides white and blue epi illumination for other types of gels and blots such as SYBR safe.
Certifications: UL, CSA and CE compliant.
Order Now
ORDER
ACCESSORIES
SERVICES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty