Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
BioX PCR Tubes and Caps
Supplier: TradeWinds (TWD)
BioX
BX60-3230 BX60-3111 BX60-3220 BX60-3102 BX60-3235 BX60-3247 BX60-3245 BX60-3240 BX60-3135
76502-782CS 1054.2 USD InStock
76502-782 76502-772 76502-774 76502-780 76502-770 76502-768 76502-776 76502-766 76502-778
BioX PCR Tubes and Caps
Tubes PCR Tubes
PCR tube and caps available individually, or in strips of 8 with domed or flat caps.
  • Convenient hinged cap design
  • Optically clear
  • Ultra thin walls
  • Positive closure feedback
All caps and tubes are certified free of detectable Rnase, Dnase, DNA, and PCR inhibitors, and are tested pyrogen-free.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty