Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
BioX PCR Plates
Supplier: TradeWinds (TWD)
BioX
BX60-3455 BX60-3441 BX60-3420 BX60-3401 BX60-3400 BX60-3425 BX60-3450 BX60-3426
76502-794CS 416.5 USD InStock
76502-794 76502-784 76502-796 76502-790 76502-792 76502-764 76502-786 76502-788
BioX PCR Plates
Microplates PCR Plates
PCR plates with broad thermal cycler compatibility.
  • High contrast indexing
  • Ultra thin walls
  • Crystal clear
  • Non sterile
  • Available with or without ridges also in skirtless, semi skirted or full skirted
All plate options are certified free of detectable Rnase, Dnase, DNA, and PCR inhibitors, and are tested pyrogen-free.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty