Would you like to log in again?
TWD BioX Microcentrifuge Tubes
Supplier: TradeWinds (TWD)
BX80-1110 BX80-1210 BX80-1310
76502-802CS 254.69 USD InStock
76502-802 76502-798 76502-800
TWD BioX Microcentrifuge Tubes
Tubes Microcentrifuge Tubes
These high quality boil proof microcentrifuge tubes are optically clear.
  • Optically clear PP
  • RNase, DNase, DNA, and PCR inhibitor free Boil proof cap
  • Positive cap closure
The crystal clear microcentrigue tubes facilitate flawless visualization of samples and are ideal for high-temp applications.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty