Would you like to log in again?
Evergreen Compartmented Biopsy Cassette Tap with Cover, Caplugs Evergreen
Supplier: Caplugs Evergreen
258-4279-W90 258-4264-W90
76502-842BG 179.4 USD InStock
76502-842 76502-844
Evergreen Compartmented Biopsy Cassette Tap with Cover, Caplugs Evergreen
Embedding Cassettes Biopsy Processing Cassettes
Caplugs Evergreen 2-, 4-, and 9-compartment cassettes with 0.6 mm holes help to customize biopsy processing with added efficiency and economy.
  • High density acetal construction
  • Acetal is resistant to histological solvents such as alcohol and xylene
  • Acetal will keep the specimen submerged
  • Lid and base preassembled
Modified ball-and-socket hinge design for ease of opening and closing and also for detachment.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty