Would you like to log in again?
Evergreen Embedding Rings, Caplugs Evergreen
Supplier: Caplugs Evergreen
258-4010-W10 258-4011-P10
76502-850CT 64.11 USD InStock
76502-850 76502-852
Evergreen Embedding Rings, Caplugs Evergreen
Embedding Cassettes Embedding Rings
Caplugs Evergreen embedding rings are for use in positioning histological tissues accurately in base molds. A treated sample is placed in the mold, the embedding ring is placed into position and the embedding medium is poured.
  • Rings are compatible with most commonly-used processing and storage systems
  • Etched labeling area is molded-in to ensure proper identification
  • Available in multiple colors
Precision-molded from premium-grade, chemically-inert, high-impact polystyrene for dimensional rigidity and sturdiness.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty