Would you like to log in again?
4-Way Collector (The Werner), SCAT
Supplier: S.C.A.T.
106660
76503-004EA 452.2 USD InStock
76503-004
4-Way Collector (The Werner), SCAT
Manifolds
This purging manifold allows you to install up to four SafetyCaps on your purging bottle.
  • Safe and easy handling - capillaries are reliably fixed
  • Solvent filters have enough space due to the wide GLS 80 opening
  • Simply close unused connections with standard bottle closures
The System for Solvent Safety stays permanently closed, the 'safety chain' remains intact during the purging process.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty