Would you like to log in again?
Accessories for Safety Funnels with Lid, 'MARCO' V2.0
Supplier: S.C.A.T.
106700 307003 318998 307101 317634 307100 307347
76948-924EA 38.03 USD InStock
76948-924 76503-024 76948-920 76948-910 76948-922 76948-912 76948-908
Accessories for Safety Funnels with Lid, 'MARCO' V2.0
Funnels Safety Funnels
Replacement parts for SCAT 'MARCO' V2.0 safety funnels, which can be closed safely and prevent the danger of ignition.
  • Suitable for all models of the MARCO series
  • Available in conductive and non-conductive versions
Stainless steel sieve to catch small objects such as stirrers from HPLC equipment.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty