Would you like to log in again?
SymLine® SwitchBox and 3-Way Ball Valve, SCAT
Supplier: S.C.A.T.
106580 160178 107000
76503-690EA 1214.99 USD InStock
76503-690 76503-028 76502-978
SymLine® SwitchBox and 3-Way Ball Valve, SCAT
Level Sensors
Automatically divert waste liquids into the reserve canister.
  • Switchbox compatible to all electronic S.C.A.T. Europe and SymLine® level sensors
  • Also compatible with the Signalbox and Built-in signalbox
  • Many more applications, not only for waste disposal of HPLC systems
Improved safety through increased collecting volume, the reserve canister protects against overflowing, even for a longer time period. For example, waste can be disposed of over the weekend without operator supervision and without having to empty or change the full container.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty