Would you like to log in again?
One-Piece Fittings with Integrated Ferrules, S.C.A.T
Supplier: S.C.A.T.
107063 107042 107061 107048 107041 107043 107059
76503-064PK 137.12 USD InStock
76503-064 76503-068 76503-066 76503-078 76503-090 76503-094 76503-092
One-Piece Fittings with Integrated Ferrules, S.C.A.T
Fittings and Connectors
These PFA / PTFE fittings connect SCAT SafetyCaps and SafetyWasteCaps to ensure that capillaries and tubes are sealed tightly.
  • Made from PFA or PTFE
  • Available in different diameters and colors
  • Autoclavable
  • High flexibility
Suitable for SafetyCaps and SafetyWasteCaps.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty