Would you like to log in again?
Accessories for Politainer, S.C.A.T.
Supplier: S.C.A.T.
107335 107338 107334 107336
76503-186EA 9.73 USD InStock
76503-186 76503-188 76503-192 76503-190
Accessories for Politainer, S.C.A.T.
Safety Cans
Cardboard boxes for SCAT Politainer for liquid waste, compatible with the SCAT system.
  • Space-saving storage
  • Easy and safe to use
  • Less expensive than comparable canisters
  • Environmentally friendly disposal
Made of chemically resistant plastic, for many applications.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty