Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Grounding Mat and Cables
Supplier: S.C.A.T.
117982 107939 108222 107938 108096 108275 108099 108011 108009 108092
76503-340PK 70.55 USD InStock
76503-340 76503-486 76503-572 76503-484 76503-342 76503-562 76503-402 76503-488 76503-400 76503-626
Grounding Mat and Cables
Grounders Grounding Wires
In order to prevent static discharge with fluids in plastic containers, the contents of the waste receptacles can be secured with ground connection. This allows users to establish an additional safety factor when collecting combustible fluids.
  • Grounding cable with different connection types
  • Cable lengths from 1.5 to 3 metres available
  • Matching with Grounding plug EU 1 megohm for connection to building services
With the different grounding cables users can safely conduct the electrostatic charge and prevent the risk of explosion due to flying sparks. The grounding cables are suitable for all SCAT and SymLine products, as well as most other suppliers.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty