Would you like to log in again?
Lab Installations for SCAT SafetyCaps, S.C.A.T.
Supplier: S.C.A.T.
108278 106502 307508 160145 107836 107886 106677 108124 106406
76503-010EA 127.27 USD InStock
76503-010 76503-574 76502-938 76503-300 76503-498 76503-682 76503-798 76502-868 76503-312
Lab Installations for SCAT SafetyCaps, S.C.A.T.
Chromatography Supplies Chromatography Spare Parts
SCAT lab installations can be completely integrated into lab furniture and offer a gastight system for safe fluid management.
SCAT lab installations fit any SCAT SafetyCap or SafetyWasteCap with GL 45 thread size, to effectively prevent health hazards and environmental damage. Changeable exhaust filter and tube connectors can be fixed directly onto the desktop for quick and easy access. The waste container can be stored under the desk, in the cupboard or sideways.
Built-in air valves and exhaust filters shut away hazardous vapors and gases, even during supply and disposal of solvents and waste fluids.
Most components are made of electroconductive HDPE and can be grounded safely.
More assembly options are available. Contact your Avantor representative for more information.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty