Would you like to log in again?
Spillage Trays, S.C.A.T.
Supplier: S.C.A.T.
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
108981 117984 117983 117986 117985
76503-592EA 344.57 USD InStock
76503-592 76503-628 76503-630 76503-634 76503-632
Spillage Trays, S.C.A.T.
Trays/Dishes/Pots Spillage Trays/Pans
Made of HDPE, white or black (electrically conductive). Containers for all purposes.
  • Electrically conductive available
  • Available with base insert; the canisters always stay dry
  • Anti-static mat and ground cable optionally available
Suitable for a wide range of canisters. Collect liquid in case of a leakage. The SCAT collection trays prevent the uncontrolled spread of hazardous liquids in the event of damage to a disposal container. The HDPE-EC conductive tray also prevents the build-up of static charge, which prevents the ignition of explosive substances.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty