Would you like to log in again?
HPLC Waste-Sets, V2.0, SCAT
Supplier: S.C.A.T.
307307 307328 307310
76503-784PK 363.84 USD InStock
76503-784 76503-766 76503-768
HPLC Waste-Sets, V2.0, SCAT
Chromatography Supplies Chromatography Spare Parts
The HPLC waste-sets, V2.0 are economical combinations for clean and safe disposal of liquid laboratory waste by HPLC systems or other analytical instruments. All sets consist of a canister and an original SCAT SafetyCap with exhaust filter as well as fittings, blind plugs and a tube connector.
  • Customised system with high-quality set components
  • Safety caps from the premium manufacturer SCAT
  • Safety canisters made of HDPE or PP material
  • Attractive price compared to single purchase
  • Ready to use, no further components necessary
SCAT offers a complete system for safe supply and disposal of solvents in analytics. All products are compatible across generations and perfectly matched to each other.
Ordering information: HPLC Waste-Set 1, V2.0: 2× 2.3/3.2 mm Ext.Ø, 1× 6.4 to 9 mm Int.Ø, filter M with exchange indicator
HPLC Waste-Set 2, V2.0: 2× 2.3/3.2 mm Ext.Ø, 1× 6.4 to 9 mm Int.Ø, filter M, with exchange indicator
HPLC Waste-Set 3, V2.0: 2× 3.2 mm Ext.Ø, 1× 5.0 to 11.5 mm Int.Ø, filter S with exchange indicator
All components can be reordered individually.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty