Would you like to log in again?
SafetyWasteSets, V2.0, SCAT
Supplier: S.C.A.T.
307316 307450 307317 307318
76503-780EA 181.33 USD InStock
76503-780 76503-794 76503-776 76503-778
SafetyWasteSets, V2.0, SCAT
Closures Safety Caps and Safety Waste Caps
We have put the most popular combinations together in our SCAT SafetyWasteSet V2.0.
  • Most popular combinations together in a complete set
  • The system is immediately ready for use
  • Comprehensive safety system in an easy to order format, simply choose which size canister is required
  • Conductivity: No
To get your waste fluid collection area ready for operation quickly, we have put the most popular combinations together in our SCAT SafetyWasteSet V2.0.
Diameter: 140 mm
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty