Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

SafetyCaps

Supplier: S.C.A.T.
399019 307410 107030 307909 307520 307006 399909 307910 107035 107032 107506 307101 107031 107742 107105 107511 107512 307019 107607 307100
76503-806EA 176.8 USD InStock
76503-806 76503-804 76503-204 76503-788 76503-986 76503-048 76503-742 76503-978 76503-802 76503-194 76503-052 76503-200 76503-750 76503-256 76948-910 76503-132 76503-198 76503-056 76948-912 76503-050
SafetyCaps
Closures Safety Caps and Safety Waste Caps
The SCAT SafetyCaps provide maximum safety for your HPLC solvent supply. Extract your solvents safely from bottles and reservoirs without being exposed to hazardous gases and solvent vapors. Complete with integrated air valve for pressure equalization.

  • High quality materials ensure maximum safety for laboratory and staff
  • 360° rotatable and up to 200 °C autoclavable screw cap for maximum flexibility
  • Stable construction for highest safety and easy handling
  • Chemically resistant against aggressive organic solvents
  • Attractive design and clear arrangement of the connections
  • Improved design for easy connection of capillaries in various colors
  • Free air vaive with change indicator included with every SafetyCap

SCAT SafetyCaps are available in the following versions:

SafetyCaps, basic: Standard SafetyCap with different connections without additional functions.
SafetyCaps, preparative: For use in the preparative HPLC environment with larger tubing connections and air valves with a higher flow rate.
SafetyCaps, with shut-off: Prevent air pockets in the tube when the supply bottle is being changed and guarantees that the analysis continues without a problem.
SafetyCaps, combined: Provides a stable baseline and reliable reproducibility for RI detection for a fast and simple solvent degassing with helium.
SafetyCaps, angled: With lateral connections for capillary and air valve in case of limited space above the bottle.
SafetyCaps, for Merck® bottles: Connect containers with thread sizes S 40 and GL 40 directly to the HPLC system, without decanting or thread adapters.
SafetyCaps, for NS 29/32 ground neck bottles: The locknut allows the stopper to be easily loosened and removed for bottles with NS 29/32 mm standard ground joints.

Delivery information: All SafetyCaps are delivered with an air valve and corresponding fittings. PFA fittings, PTFE body and PPS screw cap are conforming to the requirements of flammability classification V-0 (UL-94). Blind plugs are provided to tighten connections which are temporarily inactive or not in use.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty