Would you like to log in again?
Safety Funnels with Lid 'MARCO' V2.0, Electrically Conductive, SCAT Europe
Supplier: S.C.A.T.
318980 318981 318985 318630 318964 318960 318962 318984 317651 318961 318983 317637 306980
76503-928EA 185.68 USD InStock
76503-928 76503-926 76503-904 76503-918 76503-930 76503-920 76503-934 76503-924 76503-902 76503-932 76503-922 76503-740 76503-880
Safety Funnels with Lid 'MARCO' V2.0, Electrically Conductive, SCAT Europe
Closures Safety Caps and Safety Waste Caps
S.C.A.T. 'MARCO' V2.0 safety funnels can be closed safely and prevent danger of ignition.
  • 'MARCO', V2.0 safety lid funnel is a modern lid funnel made of high-quality HDPE with extremely high resistance to acids, alkalis and many chemicals
  • Integrated gripping edge on the funnel lid increases the grip without having to touch the possibly contoured lid
  • Innovative funnel lid with integrated sealing ring closes the funnel edge and prevents explosive or health-endangering gases escaping
  • Sturdy hinge guarantees the closing mechanism has a long service life
  • Rotating screw cap makes it easier to screw the lid funnel on and off the canister
  • With removable sieve for easy cleaning
  • The non-conductive version of the 'MARCO' V2.0 safety lid funnel is in line with the SCAT color scheme with its blue and white coloring, the conductive version is black, this difference in color helps differentiate the conductive from the non-conductive version
The innovative 'MARCO', V2.0 safety funnel is a high-quality lid funnel for the disposal of liquid waste. The lid, with an integrated rubber lip, seals the funnel and prevents the escape of toxic or explosive vapors. Numerous features guarantee safe handling. These electrically conductive versions has an earth connection to avoid ignition hazards. Available in different thread sizes for safe attachment to laboratory bottles, canisters or other containers.
Diameter: 140 mm
Delivery information: Supplied with grounding cable.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty