Would you like to log in again?
Spouts for Jerricans, S.C.A.T
Supplier: S.C.A.T.
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
610500 610501 610504 610502 610499 610503
76504-072EA 29.58 USD InStock
76504-072 76504-074 76504-064 76504-066 76504-068 76504-070
Spouts for Jerricans, S.C.A.T
Closures Safety Caps and Safety Waste Caps
For simple and safe pouring of fluids from containers.
  • Plastic
  • Suitable for S.C.A.T. safety containers
Safety Spouts are also suitable for politainer waste containers and are available in flexible and rigid versions. Designed for the common canister threads S 55, S 60/61, and S 40/71, and made of high-quality PEHD or electrically conductive PEHD-EC material with additional ventilation.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty