Would you like to log in again?
HardyCHROM™ Candida + Auris, Hardy Diagnostics
Supplier: Hardy Diagnostics
G343 G343
76509-142PK 42.89 USD InStock
CA76509-142 76509-142
HardyCHROM™ Candida + Auris, Hardy Diagnostics
Media for Microbiology Prepared Media for Microbiology
Hardy Diagnostics HardyCHROM™ Candida+auris is recommended for the selective isolation and differential identification of Candida species.
  • Candida auris produces white colonies with teal to teal-green 'bullseye' centers that are positive for UV fluorescence for easy differentiation from other Candida species
  • Colonies stand out with bright, vivid color development against a white background for effortless interpretation
  • Useful as an aid for differentiating other clinically relevant Candida species (C. albicans, C. tropicalis, C. krusei)
  • Inhibitory to most bacteria common to mixed specimens (e.g. skin, throat, axilla, urogenital tract, or rectal swabs, and more)
  • Most strains will exhibit colors within 48 hours
The medium allows for the differentiation of C. albicans, C. tropicalis, C. krusei and C. auris based on colony morphology, color, and positive UV fluorescence.
The HardyCHROM™ plates use optically clear, heavy-duty polystyrene for distortion-free viewing and reduced breakage during shipping. Receive your pack in a 'breathable' sleeve of 10 plates per bag that prevents the buildup of condensation and excess moisture.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty