Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Orum International Trio.Bas™ Remote Head, Tube and Funnel System, Hardy Diagnostics
Supplier: Hardy Diagnostics
BAS187 BAS193 BAS188 BAS199 BAS194 BAS198 BAS181 BAS195 BAS186
76509-282EA 377.14 USD InStock
76509-282 76509-272 76509-280 76509-270 76509-290 76509-268 76509-288 76509-284 76509-274
Orum International Trio.Bas™ Remote Head, Tube and Funnel System, Hardy Diagnostics
Samplers Air Samplers
This modular system is customizable according to specific equipment and microbial air sampling requirements. This remote system is particularly useful when there is limited space inside isolator cabinets or RABS.
  • Sampling system is completely fabricated in AISI 316 rated stainless steel
  • Tri-Clamp connection simplifies all cleaning/sterilization operations
  • Risk of contamination associated with operator activity is reduced
  • Remote aspirating funnel can be manipulated in multiple directions
  • Remote funnel system is fully sterilizable
Stainless steel aspirating funnel end clamp with stainless steel aspirating bell and tube end clamp to connect the system to an air sampler. All components are sold separately to allow for customization.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty