Would you like to log in again?
Boletopsis grisea Lectin (BGL), New England Biolabs
Supplier: New England Biolabs
P0867S P0867S
76509-460EA 226 USD InStock
CA76509-460 76509-460
Boletopsis grisea Lectin (BGL), New England Biolabs
Enzymes
Boletopsis grisea Lectin (BGL) is a recombinant 15 kDa lectin from the Boletopsis grisea mushroom that has been expressed in E. coli.
  • Enzyme for innovation
  • Binds to N-glycans having at least one terminal outer arm GlcNAc
  • Concentration: 1 mg/ml
Enables enrichment of GlcNAc-capped N-glycans or mucin type O-glycopeptides from complex samples in glycomics and glycoproteomics analytical workflows.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty