Would you like to log in again?
Mismatch Endonuclease I, New England Biolabs
Supplier: New England Biolabs
M0678S M0678S
76509-466EA 162 USD InStock
CA76509-466 76509-466
Mismatch Endonuclease I, New England Biolabs
Enzymes
Mismatch Endonuclease I is a Mg²⁺ dependent DNA endonuclease that specifically cleaves mismatched base pairs (T:T, G:G and T:G mismatches).
  • Recognizes and cleave T:T, G:G and T:G mismatches in DNA
  • Cleaves the 3rd phosphodiester bond on the 5' side of the mismatched base in both strands
  • Cleaves a 5 bp overhang with a 3'-OH and 5'-phosphate
  • Best at T mismatches and does not recognize all DNA mismatches
  • Isolated from a recombinant source
  • Concentration: 80000 units/ml
Mismatch Endonuclease I cleaves the 3rd phosphodiester bond on the 5' side of the mismatched base in both strands, leaving a 5 bp overhang. While Mismatch Endonuclease I prefers to cleave T:T, G:G and T:G DNA mismatches, it will also readily cleave T:I, G:I and G:U DNA mismatches. Additionally, Mismatch Endonuclease I has been shown to nick the thymine containing strand of T:C DNA mismatches, and cleave T:G and T:U (DNA:RNA) mismatches albeit to a lesser extent than DNA:DNA mismatches.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty