Would you like to log in again?
K3M Monochrome CMOS Camera, Leica Microsystems
Supplier: Leica
Leica®
11547113 11547113
76509-496EA 5335.9 USD InStock
CA76509-496 76509-496
K3M Monochrome CMOS Camera, Leica Microsystems
Cameras
For life science and industry imaging applications and analysis.
  • Very good dynamic range to acquire high quality, quantifiable images for downstream analysis
  • Large sensor and high frame rates
Recommended as a basic workhorse camera for bright fluorescent samples.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty