Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PSA Mouse Monoclonal Antibody [clone: 8A9B8]

Supplier: GenScript Biotech
GenScript®
A01664-5 A01664-1
76509-816EA 202.46 USD InStock
76509-816 76509-818
Anti-PSA Mouse Monoclonal Antibody [clone: 8A9B8]
Antibodies
PSA monoclonal antibodies (5B10D4, 8A9B8, 1G9G8, 4A5D10 and 1A7D4) recognize total PSA, freePSA and ACT- PSA Complex (Prostate Specific Antigen/a-1 Antichymotrypsin Complex).

  • High specificit
  • High affinity
  • High stability

GenScript offers highly specific monoclonal and polyclonal primary antibodies.These primary antibodies are validated for multiple applications, including Sandwich ELISA and western blot.

Type: Primary
Antigen: PSA
Clonality: Monoclonal
Clone: 8A9B8
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty