Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Camelid VHH Rabbit Monoclonal Antibody [clone: 96A3F5] (Biotin)

Supplier: GenScript Biotech
GenScript®
A01995-200
76510-014EA 224.3 USD InStock
76510-014
Anti-Camelid VHH Rabbit Monoclonal Antibody [clone: 96A3F5] (Biotin)
Antibodies
GenScript MonoRab Rabbit Anti-Camelid VHH Antibody [Biotin], mAb is specific for Camelid VHH Antibody (Alpaca, Llama and Camel). The monoclonal antibody has no cross-reactivity with mouse, rat, rabbit, chicken, goat or human immunoglobulins.

  • Anti-VHH Antibodies
  • MonoRab™ technology guarantees high affinity: Kd ~10 to 11 M
  • No cross reactivity with other species
  • Specific to camelid IgG2 and IgG3
  • Recognize conformational epitopes of the VHH domain
  • Recognize both camelid and humanized VHHs

GenScript has developed a series of rabbit monoclonal (MonoRab) anti-VHH antibodies to facilitate your research of VHHs. Our anti-VHH antibodies recognize conformational epitopes of the VHH domain of camelid lgG2 and lgG3 subclasses. It is not necessary to add a tag to VHH for detection anymore.

Type: Primary
Antigen: Camelid VHH
Clonality: Monoclonal
Clone: 96A3F5
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Alpaca;Llama;Camel

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty