Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgG Rabbit Monoclonal Antibody [clone: 37C2]

Supplier: Genscript
GenScript®
A02022-200
76510-060EA 115.2 USD InStock
76510-060
Anti-IgG Rabbit Monoclonal Antibody [clone: 37C2]
Antibodies
Rabbit IgG control is suitable for use as control, standard, blocking agent, or coating protein in a variety of assays, including ELISA, western blot, immunoprecipitation, immunodiffusion, immunoelectrophoresis, flow cytometry and immunofluorescence. GenScript MonoRab™ Rabbit IgG Control (whole molecule), mAb is a rabbit monoclonal antibody, highly purified by protein A affinity column from cell culture in vitro under conditions free from animal-derived components.

  • High specificit
  • High affinity
  • High stability

GenScript offers a wide selection of polyclonal and monoclonal loading control antibodies for α-tubulin and β-actin for several species. HRP- and biotin-conjugated versions of these antibodies are also available. GenScript also offers a series of isotype control antibodies that can be used in your research.
.

Type: Control 
Antigen: IgG
Clonality: Monoclonal
Clone: 37C2
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rabbit

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty