Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SARS-CoV-2 Spike S1 Human Monoclonal Antibody [clone: HC2001]

Supplier: GenScript Biotech
GenScript®
A02038-100 A02038-1
76510-092EA 3135 USD InStock
76510-092 76510-094
Anti-SARS-CoV-2 Spike S1 Human Monoclonal Antibody [clone: HC2001]
Antibodies
The product is specific for SARS-CoV-2 Spike Protein S1 subunit and its RBD domain.

  • COVID-19 antibodies
  • Validated for ELISA, WB, FACS and SARS-CoV-2 virus neutralization assay
  • Control antibodies and various antibody pairs available for ELISA-based detection
  • Neutralizing antibody standard to calibrate quantitative assay of SARS-CoV-2 neutralizing antibodies in samples

Genscript offers site-specific neutralizing antibodies, neutralizing antibody standards and control antibodies.

Type: Primary
Antigen: SARS-CoV-2 Spike S1
Clonality: Monoclonal
Clone: HC2001
Conjugation: Unconjugated
Public Immunogen Range:
Host: Human
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty