Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SARS-CoV-2 RBD Mouse Monoclonal Antibody [clone: 5B7D7]

Supplier: Genscript
GenScript®
A02056-1 A02056-100
76510-150EA 3135 USD InStock
76510-150 76510-152
Anti-SARS-CoV-2 RBD Mouse Monoclonal Antibody [clone: 5B7D7]
Antibodies
The product is specific for SARS-CoV-2 spike protein S1 subunit and its RBD domain.

  • COVID-19 antibodies
  • Validated for ELISA, WB, FACS, and SARS-CoV-2 virus neutralization assay
  • Control antibodies and various antibody pairs available for ELISA based detection
  • Neutralizing antibody standard to calibrate quantitative assay of SARS-CoV-2 neutralizing antibodies in samples

Genscript offers site-specific neutralizing antibodies, neutralizing antibody standards, and control antibodies.

Type: Primary
Antigen: SARS-CoV-2 RBD
Clonality: Monoclonal
Clone: 5B7D7
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty