Would you like to log in again?
KIT HIGH-STABILITY PCR
Supplier: Genscript
High-Stability PCR Kit contains Green Taq DNA polymerase, 10x Taq buffer, and Stabilized dNTP Mix, 10 mM Each, all specially formulated for increased stability using our proprietary technology. The kit can be stored at 4℃. For over six months or at room temperature (25℃) for one mon
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
KIT HIGH-STABILITY PCR 76510-300 L00342 Each Retrieving
Temperature sensitive Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty