Would you like to log in again?
eBlot™ L1 Fast Wet Transfer System
Supplier: Genscript
eBlot™
L00686
76510-444EA 5947.55 USD InStock
76510-444
eBlot™ L1 Fast Wet Transfer System
Blotting Apparatus
eBlot™ L1 wet protein transfer system is a fast western blotting protein transfer system that combines the high reproducibility and efficiency of the traditional wet protein transfer with the speed and convenience of semi-dry protein transfer. It allows efficient western blot transfer of small, medium, and large molecular weight proteins.
  • Automated fast wet protein transfer: 1 or 2 mini gels in 9 to 17 minutes
  • Full protein size coverage: Outstanding performance for small, medium, and large molecular weight proteins
  • High protein transfer efficiency: With gold-standard wet protein transfer technology
  • Convenience: Unique cassette sandwich setup allows easy and clean operation
  • Flexibility: Customizable program
  • All-inclusive consumable kit: Include precut NC/PVDF membranes and buffers with a long shelf life
  • Highly compatible with different types of precast or homemade mini gels
eBlot™ L1 allows fast and efficient protein transfer of 1 or 2 mini gels at a time. The system comes with conveniently designed transfer stacks/consumables (sold separately). The setup takes less than 2 minutes and can be performed even with the sponge in a dry state (patented).
Why Choose eBlot™ L1? Conventional western blot wet transfers are cumbersome and require experience and a great deal of time spent on consumable preparation. The transfer times usually vary from one hour to overnight. With eBlot™ L1 fast wet protein transfer system, achieve excellent western blot transfer efficiency without sacrificing speed.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty