Would you like to log in again?
HRP-PROTEIN G 1MG
Supplier: Genscript
Protein G, a cell surface protein of Group G streptococci,Type III Fc receptor that binds to Fc region of IgG by a non-immune mechanism similar to that of protein A. It contains 3 IgG binding domains & is engineered to delete albumin-binding region. HRP has been covalently coupled to Pro, Size: 1mg
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
HRP-PROTEIN G 1MG 76510-530 M00090 Each (1mg) Retrieving
Temperature sensitive Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty