Would you like to log in again?
HRP-PROTEIN L 1MG
Supplier: Genscript
Protein L is a cell surface protein expressed by Peptostreptoccocus magnus, that binds to variable light chains (kappa chains) of immunoglobulins without interfering with antigen binding. In contrast to IgG-binding proteins such as protein A and protein G, protein L can be used for d, Size: 1mg
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
HRP-PROTEIN L 1MG 76510-538 M00098 Each (1mg) Retrieving
Temperature sensitive Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty